Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr. 12: 81860028 - 82000924 on Build GRCm38
UniGene ID
Mm.86584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018814.3 NP_061284.2 15 3764
GenBank mRNA AF096286.1 - 9 1594
AK129221.1 - 10 1644
BC057333.1 - 8 1507
BC098370.1 - 10 1354

View Full Product Details