Gene Transcripts

Gene Symbol
SF3A3
Entrez Gene ID
10946
Gene Name
splicing factor 3a subunit 3
Gene Aliases
PRP9, PRPF9, SAP61, SF3a60
Chromosome Location
Chr. 1: 37956975 - 37990110 on Build GRCh38
UniGene ID
Hs.77897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006802.3 NP_006793.1 3 326
GenBank mRNA AK128438.1 - 3 1135
AK222554.1 - 3 202
AK291181.1 - 3 207
BC002395.2 - 3 219 2821350
BC011523.1 - 3 283 3867442
DA835019.1 - 3 326
U08815.1 - 3 189

View Full Product Details