Gene Transcripts

Gene Symbol
Cts7
Entrez Gene ID
56092
Gene Name
cathepsin 7
Gene Aliases
AA408230, AA408952, CTS1, Epcs24, Epcs71
Chromosome Location
Chr. 13: 61352461 - 61358290 on Build GRCm38
UniGene ID
Mm.3692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019539.3 NP_062412.1 4 511
GenBank mRNA AF250837.1 - 3 298
AY014779.1 - 4 371
BC050198.1 - 3 607
BC064740.1 - 4 511 30018572

View Full Product Details