Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019635.2 | NP_062609.2 | 9, 10 | 1408 | |
GenBank mRNA | AF271361.1 | - | 9, 10 | 1154 | |
AK002914.1 | - | 4, 5 | 626 | ||
AY058922.1 | - | 9, 10 | 1294 | ||
BC037440.1 | - | 7, 8 | 1164 | 3585312 | |
BC049123.1 | - | 9, 10 | 1282 | 6813956 | |
U28726.1 | - | 9, 10 | 1287 |