Gene Transcripts

Gene Symbol
MORF4L1
Entrez Gene ID
10933
Gene Name
mortality factor 4 like 1
Gene Aliases
Eaf3, FWP006, HsT17725, MEAF3, MORFRG15, MRG15, S863-6
Chromosome Location
Chr. 15: 78872781 - 78897739 on Build GRCh38
UniGene ID
Hs.374503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001265604.1 NP_001252533.1 3 306
NM_001265605.1 NP_001252534.1 3 206
NM_006791.3 NP_006782.1 3 353
NM_206839.2 NP_996670.1 3 353
XM_017021872.1 XP_016877361.1 3 318
GenBank mRNA AF070664.1 - 3 160
AF100615.1 - 3 247
AF109188.1 - 3 142
AF161546.1 - 3 139
AF167173.1 - 3 155
AF218011.1 - 3 258
AK095823.1 - 4 755
AK098485.1 - 3 273
AK296650.1 - 3 244
AK300789.1 - 3 290
AK310181.1 - 2 91
AK310592.1 - 3 152
AK310702.1 - 3 161
AL137697.1 - 3 147
AY148481.1 - 3 247
BC002936.2 - 3 234 3955120
BC022845.2 - 3 234 2988803
BC067826.1 - 3 129 4824022
BC141840.1 - 3 161 8860303
BG771854.1 - 3 363
BX382045.2 - 3 150
DB455429.1 - 3 206

View Full Product Details