Gene Transcripts

Gene Symbol
Txnl4a
Entrez Gene ID
27366
Gene Name
thioredoxin-like 4A
Gene Aliases
D18Wsu98e, Dim1, ENSMUSG00000057130, Txnl4, U5-15kD, U5-15kDa
Chromosome Location
Chr. 18: 80206794 - 80225851 on Build GRCm38
UniGene ID
Mm.172411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038608.2 NP_001033697.2 4 1585
NM_001042408.1 NP_001035867.1 4 1561
NM_178604.3 NP_848719.2 4 1697
GenBank mRNA AK010599.1 - 4 1585
AK011717.1 - 4 1649
AK012775.1 - 1 511
AK171121.1 - 4 1531
BC003876.1 - 4 1503
BC014770.1 - 4 1561
BC025053.1 - 4 1531
BC029327.1 - 4 1548

View Full Product Details