Gene Transcripts

Gene Symbol
Zfp61
Entrez Gene ID
22719
Gene Name
zinc finger protein 61
Gene Aliases
Chromosome Location
Chr. 7: 24291045 - 24299549 on Build GRCm38
UniGene ID
Mm.12832
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009561.2 NP_033587.2 7 1421
GenBank mRNA AK148158.1 - 7 1239
AK149960.1 - 7 1216
BC006618.1 - 7 1460 3967527

View Full Product Details