Gene Transcripts

Gene Symbol
Trap1a
Entrez Gene ID
22037
Gene Name
tumor rejection antigen P1A
Gene Aliases
Tra-P1A
Chromosome Location
Chr. X: 139333683 - 139338165 on Build GRCm38
UniGene ID
Mm.1297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011635.1 NP_035765.1 1 636
GenBank mRNA AK146737.1 - 1 451
BC051644.1 - 1 429 30022193
M36387.1 - 1 636

View Full Product Details