Gene Transcripts
- Gene Symbol
- Tnfrsf4
- Entrez Gene ID
- 22163
- Gene Name
- tumor necrosis factor receptor superfamily, member 4
- Gene Aliases
- ACT35, CD134, Ly-70, Ox40, TXGP1L, Txgp1
- Chromosome Location
- Chr. 4: 156013695 - 156016613 on Build GRCm38
- UniGene ID
- Mm.13885
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z21674.1 |
- |
7 |
828 |
|
View Full Product Details