Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110328.1 | NP_001103798.1 | 3 | 1861 | |
NM_001110329.1 | NP_001103799.1 | 3 | 1804 | ||
NM_001110330.1 | NP_001103800.1 | 3 | 1734 | ||
NM_011813.3 | NP_035943.3 | 3 | 1845 | ||
GenBank mRNA | AF126746.1 | - | 3 | 1795 | |
AF126747.1 | - | 3 | 1803 | ||
AK003641.1 | - | 1 | 955 | ||
AK135672.1 | - | 3 | 1735 | ||
AK142205.1 | - | 3 | 1754 | ||
AK162454.1 | - | 3 | 1726 | ||
AK170633.1 | - | 3 | 1730 | ||
AK177756.1 | - | 1 | 150 | ||
AK182118.1 | - | 1 | 146 | ||
AK220212.1 | - | 3 | 1734 | ||
BC006633.1 | - | 3 | 1845 | 3482191 | |
BC106113.1 | - | 3 | 1720 | 6492060 | |
BC138231.1 | - | 3 | 1795 | 8861250 | |
BC138232.1 | - | 3 | 1820 | 8861251 | |
BC145207.1 | - | 3 | 1820 | 9053758 |