Gene Transcripts

Gene Symbol
Tenm1
Entrez Gene ID
23963
Gene Name
teneurin transmembrane protein 1
Gene Aliases
Odz1, Odz2, TCAP-1, Ten-m1, ten-1
Chromosome Location
Chr. X: 42527866 - 43429156 on Build GRCm38
UniGene ID
Mm.327698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011855.4 NP_035985.2 16 2705
GenBank mRNA AB025410.1 - 16 2707
BC138860.1 - 15 2688 8861882
BC138861.1 - 15 2688 8861883

View Full Product Details