Gene Transcripts

Gene Symbol
ZNF19
Entrez Gene ID
7567
Gene Name
zinc finger protein 19
Gene Aliases
KOX12
Chromosome Location
Chr. 16: 71474073 - 71489351 on Build GRCh38
UniGene ID
Hs.660076
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006961.3 NP_008892.2 6 592
GenBank mRNA AK055567.1 - 5 473
AK057330.1 - 5 767
AK292260.1 - 6 790
BC047091.1 - 6 579 5270853

View Full Product Details