Gene Transcripts
- Gene Symbol
- Yme1l1
- Entrez Gene ID
- 27377
- Gene Name
- YME1-like 1 (S. cerevisiae)
- Gene Aliases
- FtsH1, Ftsh
- Chromosome Location
- Chr. 2: 23156505 - 23199260 on Build GRCm38
- UniGene ID
- Mm.23335
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013771.5 |
NP_038799.1
|
2, 3 |
367 |
|
GenBank mRNA |
AF090430.1 |
- |
2, 3 |
345 |
|
AK032273.1 |
- |
2, 3 |
355 |
|
AK045141.1 |
- |
2, 3 |
352 |
|
AK079175.1 |
- |
2, 3 |
327 |
|
AK137665.1 |
- |
2, 3 |
319 |
|
AY136286.1 |
- |
2, 3 |
183 |
|
AY136287.1 |
- |
2, 3 |
336 |
|
BB624412.1 |
- |
2, 3 |
367 |
|
BC007128.1 |
- |
2, 3 |
328 |
3591123 |
View Full Product Details