Gene Transcripts

Gene Symbol
Nr5a1
Entrez Gene ID
26423
Gene Name
nuclear receptor subfamily 5, group A, member 1
Gene Aliases
Ad4BP, ELP, ELP-3, Ftz-F1, Ftzf1, SF-1, SF1
Chromosome Location
Chr. 2: 38692656 - 38714560 on Build GRCm38
UniGene ID
Mm.31387
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139051.3 NP_620639.1 7 1959
GenBank mRNA AK134378.1 - 7 1960
AK144376.1 - 7 1921
AK156911.1 - 7 1927

View Full Product Details