Gene Transcripts

Gene Symbol
Klk6
Entrez Gene ID
29245
Gene Name
kallikrein related-peptidase 6
Gene Aliases
Prss9
Chromosome Location
Chr. 1: 99762221 - 99769496 on Build Rnor_6.0
UniGene ID
Rn.10732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019175.1 NP_062048.1 5, 6 657
XM_006228982.2 XP_006229044.1 5, 6 764
GenBank mRNA AF016269.1 - 5, 6 657

View Full Product Details