Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007023.3 | NP_008954.2 | 3 | 414 | |
XM_005246246.2 | XP_005246303.1 | 3 | 281 | ||
XM_006712205.3 | XP_006712268.1 | 3 | 286 | ||
XM_011510505.2 | XP_011508807.1 | 3 | 281 | ||
XM_017003196.1 | XP_016858685.1 | 3 | 281 | ||
GenBank mRNA | AB027471.1 | - | 3 | 284 | |
AB209681.1 | - | 3 | 343 | ||
AK294997.1 | - | 3 | 379 | ||
AK310373.1 | - | 3 | 451 | ||
AK313084.1 | - | 3 | 283 | ||
BC024004.2 | - | 3 | 253 | 4823935 | |
KJ534924.1 | - | 3 | 191 | ||
KJ534925.1 | - | 3 | 227 | ||
KJ534978.1 | - | 3 | 227 | ||
U78516.1 | - | 3 | 335 |