Gene Transcripts

Gene Symbol
Gng11
Entrez Gene ID
64199
Gene Name
guanine nucleotide binding protein (G protein), gamma 11
Gene Aliases
Chromosome Location
Chr. 4: 28989170 - 28994610 on Build Rnor_6.0
UniGene ID
Rn.892
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022396.2 NP_071791.1 2 681
GenBank mRNA AF257110.1 - 2 401
BC097320.1 - 2 681 7455035

Gene Symbol
LOC100912034
Entrez Gene ID
100912034
Gene Name
guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11-like
Gene Aliases
Chromosome Location
Chr. 4: 31387262 - 31392791 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749698.3 XP_003749746.1 2 839

View Full Product Details