Gene Transcripts

Gene Symbol
Vax2
Entrez Gene ID
64572
Gene Name
ventral anterior homeobox 2
Gene Aliases
Chromosome Location
Chr. 4: 115388839 - 115412515 on Build Rnor_6.0
UniGene ID
Rn.48765
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022637.1 NP_072159.1 2 268
GenBank mRNA AF113516.1 - 2 268

View Full Product Details