Gene Transcripts

Gene Symbol
Kcnh2
Entrez Gene ID
117018
Gene Name
potassium channel, voltage gated eag related subfamily H, member 2
Gene Aliases
ERG1
Chromosome Location
Chr. 4: 7355161 - 7387243 on Build Rnor_6.0
UniGene ID
Rn.10970
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053949.1 NP_446401.1 3 432
XM_006235867.2 XP_006235929.1 3 565
GenBank mRNA Z96106.1 - 3 432

View Full Product Details