Gene Transcripts

Gene Symbol
Rps23
Entrez Gene ID
124323
Gene Name
ribosomal protein S23
Gene Aliases
Chromosome Location
Chr. 2: 19823234 - 19824804 on Build Rnor_6.0
UniGene ID
Rn.2139
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_078617.3 NP_511172.1 2 185
GenBank mRNA BC058134.1 - 2 189

View Full Product Details