Gene Transcripts

Gene Symbol
Pde7b
Entrez Gene ID
140929
Gene Name
phosphodiesterase 7B
Gene Aliases
Chromosome Location
Chr. 1: 15889845 - 16203909 on Build Rnor_6.0
UniGene ID
Rn.234527
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080894.2 NP_543170.1 8 766
XM_008758591.1 XP_008756813.1 7 1086
XM_008758592.1 XP_008756814.1 7 670
GenBank mRNA AB057409.1 - 8 766
AB057410.1 - 7 634
AB057411.1 - 9 735

View Full Product Details