Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_147211.1 | NP_671744.1 | 7 | 1500 | |
XM_008762874.1 | XP_008761096.1 | 8 | 2037 | ||
XM_008762875.1 | XP_008761097.1 | 8 | 4364 | ||
XM_008762876.1 | XP_008761098.1 | 8 | 2037 | ||
XM_008762877.1 | XP_008761099.1 | 8 | 2035 | ||
GenBank mRNA | U25281.1 | - | 7 | 1500 | |
U31159.1 | - | 8 | 1602 |