Gene Transcripts

Gene Symbol
Eif2b1
Entrez Gene ID
64514
Gene Name
eukaryotic translation initiation factor 2B, subunit 1 alpha
Gene Aliases
eIF-2a
Chromosome Location
Chr. 12: 37444136 - 37452391 on Build Rnor_6.0
UniGene ID
Rn.9181
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172029.2 NP_742026.1 3 202
GenBank mRNA BC081709.1 - 3 202
FQ217438.1 - 3 240
U05821.1 - 3 221

View Full Product Details