Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 4 | 501 | |
GenBank mRNA | AF021351.1 | - | 5 | 368 | |
AK300329.1 | - | 4 | 481 | ||
AY091459.1 | - | 4 | 368 | ||
BC041089.1 | - | 4 | 462 | 3915501 | |
DA103416.1 | - | 4 | 501 |