Gene Transcripts

Gene Symbol
Zar1
Entrez Gene ID
353228
Gene Name
zygote arrest 1
Gene Aliases
Chromosome Location
Chr. 14: 37759776 - 37763712 on Build Rnor_6.0
UniGene ID
Rn.96129
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181385.2 NP_852050.1 3, 4 962
GenBank mRNA AY283175.1 - 3, 4 962

View Full Product Details