Gene Transcripts

Gene Symbol
Map1lc3b
Entrez Gene ID
64862
Gene Name
microtubule-associated protein 1 light chain 3 beta
Gene Aliases
Map1lc3, Mpl3, zbs559
Chromosome Location
Chr. 19: 53635633 - 53643965 on Build Rnor_6.0
UniGene ID
Rn.41412
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022867.2 NP_074058.2 4 1090
GenBank mRNA AY206670.1 - 1 148
AY392036.1 - 4 1090
BC083556.1 - 4 1070 7189855

View Full Product Details