Gene Transcripts

Gene Symbol
Mtr
Entrez Gene ID
81522
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase
Gene Aliases
methioninesynthase
Chromosome Location
Chr. 17: 66210444 - 66295014 on Build Rnor_6.0
UniGene ID
Rn.205061
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030864.1 NP_110491.1 2 368
GenBank mRNA AF034214.1 - 2 368

View Full Product Details