Gene Transcripts

Gene Symbol
Arid4b
Entrez Gene ID
84481
Gene Name
AT rich interactive domain 4B (Rbp1 like)
Gene Aliases
Bcaa
Chromosome Location
Chr. 17: 53831353 - 53955920 on Build Rnor_6.0
UniGene ID
Rn.47249
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053421.1 NP_445873.1 2, 3 163
XM_006254016.2 XP_006254078.1 3, 4 488
XM_006254019.2 XP_006254081.1 3, 4 488
XM_006254020.2 XP_006254082.1 3, 4 488
XM_008771764.1 XP_008769986.1 3, 4 488
XM_008771765.1 XP_008769987.1 3, 4 488
XM_008771766.1 XP_008769988.1 3, 4 488
GenBank mRNA AF245512.1 - 2, 3 163

Gene Symbol
LOC100912163
Entrez Gene ID
100912163
Gene Name
AT-rich interactive domain-containing protein 4B-like
Gene Aliases
Chromosome Location
Chr. 17: 53629443 - 53741761 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771836.1 XP_008770058.1 2, 3 248

View Full Product Details