Gene Transcripts

Gene Symbol
Hacl1
Entrez Gene ID
85255
Gene Name
2-hydroxyacyl-CoA lyase 1
Gene Aliases
Hpcl2, Phyh2
Chromosome Location
Chr. 16: 7720047 - 7758119 on Build Rnor_6.0
UniGene ID
Rn.219233
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053493.1 NP_445945.1 6 550
GenBank mRNA AJ517469.1 - 6 550
BC078697.1 - 6 534 7110355

View Full Product Details