Gene Transcripts

Gene Symbol
UST4r
Entrez Gene ID
171397
Gene Name
integral membrane transport protein UST4r
Gene Aliases
Chromosome Location
Chr. 1: 224319927 - 224389499 on Build Rnor_6.0
UniGene ID
Rn.81213
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134379.1 NP_599206.1 6 1172
GenBank mRNA AJ132859.1 - 6 1172
BC100065.1 - 1062 7373541
FQ210050.1 - 1093
FQ218143.1 - 1232

View Full Product Details