Gene Transcripts

Gene Symbol
NEWGENE_619909
Entrez Gene ID
103690034
Gene Name
HAUS augmin-like complex, subunit 1
Gene Aliases
Haus1
Chromosome Location
Chr. 18: 74138879 - 74151990 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772211.1 XP_008770433.1 2500

Gene Symbol
Haus1
Entrez Gene ID
192228
Gene Name
HAUS augmin-like complex, subunit 1
Gene Aliases
Ccdc5
Chromosome Location
Chr. 18: 74187161 - 74198369 on Build Rnor_6.0
UniGene ID
Rn.163388
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF092207.1 - 2469

View Full Product Details