Gene Transcripts

Gene Symbol
Rbm45
Entrez Gene ID
266631
Gene Name
RNA binding motif protein 45
Gene Aliases
Drb1
Chromosome Location
Chr. 3: 63220660 - 63235638 on Build Rnor_6.0
UniGene ID
Rn.204106
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153306.1 NP_695218.1 1 156
GenBank mRNA AB036990.1 - 1 156

View Full Product Details