Gene Transcripts

Gene Symbol
PNMA2
Entrez Gene ID
10687
Gene Name
paraneoplastic Ma antigen 2
Gene Aliases
MA2, MM2, RGAG2
Chromosome Location
Chr. 8: 26504680 - 26513967 on Build GRCh38
UniGene ID
Hs.591838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007257.5 NP_009188.1 3 898
XM_011544365.2 XP_011542667.1 3 997
GenBank mRNA AB020690.1 - 1 305
AF037365.1 - 1 123
AK055325.1 - 3 840
BC036489.1 - 3 828 5262276
BC047515.1 - 3 803 5294654
BC062301.1 - 1 379 6580976

View Full Product Details