Gene Transcripts

Gene Symbol
SYNRG
Entrez Gene ID
11276
Gene Name
synergin, gamma
Gene Aliases
AP1GBP1, SYNG
Chromosome Location
Chr. 17: 37514797 - 37613637 on Build GRCh38
UniGene ID
Hs.594647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163544.2 NP_001157016.1 13 2165
NM_001163545.2 NP_001157017.1 13 2162
NM_001163546.2 NP_001157018.1 13 2165
NM_001163547.2 NP_001157019.1 12 1916
NM_007247.5 NP_009178.3 14 2399
NM_080550.4 NP_542117.3 13 2165
NM_198882.2 NP_942583.1 13 2165
XM_005256980.4 XP_005257037.1 15 2760
XM_017024076.1 XP_016879565.1 15 2760
XM_017024077.1 XP_016879566.1 15 2679
XM_017024078.1 XP_016879567.1 15 2760
XM_017024079.1 XP_016879568.1 15 2760
XM_017024080.1 XP_016879569.1 15 6812
XM_017024081.1 XP_016879570.1 15 6809
XM_017024082.1 XP_016879571.1 15 2760
XM_017024083.1 XP_016879572.1 15 2760
XM_017024084.1 XP_016879573.1 15 2760
XM_017024085.1 XP_016879574.1 15 2760
XM_017024086.1 XP_016879575.1 15 2760
XM_017024087.1 XP_016879576.1 15 2760
XM_017024088.1 XP_016879577.1 14 2526
XM_017024089.1 XP_016879578.1 15 2760
XM_017024090.1 XP_016879579.1 14 2457
XM_017024091.1 XP_016879580.1 14 2376
XM_017024092.1 XP_016879581.1 15 2760
XM_017024093.1 XP_016879582.1 14 2457
XM_017024094.1 XP_016879583.1 14 2376
XM_017024095.1 XP_016879584.1 14 2457
XM_017024096.1 XP_016879585.1 15 2760
XM_017024097.1 XP_016879586.1 14 2457
XM_017024098.1 XP_016879587.1 14 2457
XM_017024099.1 XP_016879588.1 13 2142
XM_017024100.1 XP_016879589.1 14 2457
XM_017024101.1 XP_016879590.1 14 2376
XM_017024102.1 XP_016879591.1 13 2142
XM_017024103.1 XP_016879592.1 13 2223
XM_017024104.1 XP_016879593.1 13 2223
XM_017024106.1 XP_016879595.1 13 2223
GenBank mRNA AF169548.1 - 14 2359
AK025335.1 - 7 1555
AK025700.1 - 1 412
BC090930.1 - 13 2116 3077104
BC117313.1 - 13 2105 40125864
BC143476.1 - 12 1859 9051984
BC143478.1 - 13 2108 9051986

View Full Product Details