Gene Transcripts

Gene Symbol
Pglyrp1
Entrez Gene ID
84387
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
Pglyrp, Pgrp
Chromosome Location
Chr. 1: 79790879 - 79796064 on Build Rnor_6.0
UniGene ID
Rn.2834
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053373.1 NP_445825.1 3 564
GenBank mRNA AF154114.1 - 3 564

View Full Product Details