Gene Transcripts

Gene Symbol
Bbs2
Entrez Gene ID
113948
Gene Name
Bardet-Biedl syndrome 2
Gene Aliases
Chromosome Location
Chr. 19: 11385865 - 11421517 on Build Rnor_6.0
UniGene ID
Rn.15987
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053618.1 NP_446070.1 1, 2 215
XM_006255131.2 XP_006255193.1 1, 2 374
GenBank mRNA AF342738.1 - 1, 2 215

View Full Product Details