Gene Transcripts

Gene Symbol
Stx1a
Entrez Gene ID
116470
Gene Name
syntaxin 1A (brain)
Gene Aliases
Chromosome Location
Chr. 12: 24682046 - 24710047 on Build Rnor_6.0
UniGene ID
Rn.9943
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053788.2 NP_446240.2 4 295
GenBank mRNA AF217191.1 - 4 264
D10392.1 - 4 295
D12519.1 - 4 255
M95734.1 - 4 256

View Full Product Details