Gene Transcripts
- Gene Symbol
- Sstr5
- Entrez Gene ID
- 25354
- Gene Name
- somatostatin receptor 5
- Gene Aliases
- Smstr5, Somato
- Chromosome Location
- Chr. 10: 14847749 - 14853827 on Build Rnor_6.0
- UniGene ID
- Rn.91342
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012882.2 |
NP_037014.2
|
2 |
769 |
|
GenBank mRNA |
L04535.1 |
- |
2 |
772 |
|
View Full Product Details