Gene Transcripts

Gene Symbol
Ccng1
Entrez Gene ID
25405
Gene Name
cyclin G1
Gene Aliases
CYCG, Ccng
Chromosome Location
Chr. 10: 25903925 - 25910298 on Build Rnor_6.0
UniGene ID
Rn.5834
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012923.2 NP_037055.1 5 876
GenBank mRNA BC081852.1 - 5 847
FQ211577.1 - 4 849
FQ211975.1 - 5 928
FQ224502.1 - 5 837
X70871.1 - 5 846

View Full Product Details