Gene Transcripts

Gene Symbol
Cask
Entrez Gene ID
29647
Gene Name
calcium/calmodulin-dependent serine protein kinase (MAGUK family)
Gene Aliases
Chromosome Location
Chr. X: 9815652 - 10156155 on Build Rnor_6.0
UniGene ID
Rn.72627
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022184.1 NP_071520.1 3 545
GenBank mRNA U47110.1 - 3 545
Y08769.1 - 43

View Full Product Details