Gene Transcripts

Gene Symbol
ACOT9
Entrez Gene ID
23597
Gene Name
acyl-CoA thioesterase 9
Gene Aliases
ACATE2, CGI-16, MT-ACT48, MTACT48
Chromosome Location
Chr. X: 23701055 - 23743290 on Build GRCh38
UniGene ID
Hs.298885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033583.2 NP_001028755.2 5 498
NM_001037171.1 NP_001032248.1 6 525
XM_005274472.2 XP_005274529.1 4 397
GenBank mRNA AF132950.1 - 4 400
AK000659.1 - 5 484
AK024337.1 - 7 1590
AL548456.3 - 6 525
BC012573.1 - 5 469
BC136671.1 - 5 449 9020661
BC144360.1 - 5 449 9052881
BC144370.1 - 5 449
BX648512.1 - 6 509

View Full Product Details