Gene Transcripts

Gene Symbol
Slc9a1
Entrez Gene ID
24782
Gene Name
solute carrier family 9, subfamily A (NHE1, cation proton antiporter 1), member 1
Gene Aliases
Nhe1
Chromosome Location
Chr. 5: 151573122 - 151626360 on Build Rnor_6.0
UniGene ID
Rn.5025
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012652.1 NP_036784.1 1 1030
GenBank mRNA M85299.1 - 1 1030

View Full Product Details