Gene Transcripts
- Gene Symbol
- Hk1
- Entrez Gene ID
- 25058
- Gene Name
- hexokinase 1
- Gene Aliases
- HEXOKIN
- Chromosome Location
- Chr. 20: 31911460 - 31979780 on Build Rnor_6.0
- UniGene ID
- Rn.11017
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012734.1 |
NP_036866.1
|
3, 4 |
474 |
|
GenBank mRNA |
J04526.1 |
- |
3, 4 |
474 |
|
View Full Product Details