Gene Transcripts

Gene Symbol
Itga1
Entrez Gene ID
25118
Gene Name
integrin, alpha 1
Gene Aliases
Chromosome Location
Chr. 2: 47120340 - 47281387 on Build Rnor_6.0
UniGene ID
Rn.91044
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030994.2 NP_112256.1 7 1072
XM_008760735.1 XP_008758957.1 7 688
GenBank mRNA X52140.1 - 7 1096

View Full Product Details