Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012465.3 | NP_036597.1 | 3, 4 | 600 | |
GenBank mRNA | AB023149.2 | - | 3, 4 | 550 | |
AF059516.1 | - | 3, 4 | 723 | ||
BC013871.2 | - | 3, 4 | 560 | ||
BC112341.1 | - | 3, 4 | 483 | 40038720 | |
BC112366.1 | - | 3, 4 | 483 | 40038726 | |
BC113577.1 | - | 3, 4 | 416 | 8322629 |