Gene Transcripts

Gene Symbol
Brinp2
Entrez Gene ID
286895
Gene Name
bone morphogenetic protein/retinoic acid inducible neural-specific 2
Gene Aliases
Fam5b
Chromosome Location
Chr. 13: 75948679 - 76049363 on Build Rnor_6.0
UniGene ID
Rn.22596
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173115.1 NP_775138.1 3 1295
GenBank mRNA AB077853.1 - 3 1295

View Full Product Details