Gene Transcripts

Gene Symbol
Ns5atp9
Entrez Gene ID
300795
Gene Name
NS5A (hepatitis C virus) transactivated protein 9
Gene Aliases
PAF15, Paf
Chromosome Location
Chr. 8: 71514281 - 71526182 on Build Rnor_6.0
UniGene ID
Rn.7233
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201418.1 NP_958821.1 2 97
GenBank mRNA AY496944.1 - 2 97

View Full Product Details