Gene Transcripts
- Gene Symbol
- SPO11
- Entrez Gene ID
- 23626
- Gene Name
- SPO11, initiator of meiotic double stranded breaks
- Gene Aliases
- CT35, SPATA43, TOPVIA
- Chromosome Location
- Chr. 20: 57329757 - 57343993 on Build GRCh38
- UniGene ID
- Hs.159737
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012444.2 |
NP_036576.1
|
4, 5 |
502 |
|
NM_198265.1 |
NP_937998.1
|
3, 4 |
388 |
|
XM_005260379.3 |
XP_005260436.1
|
4, 5 |
520 |
|
XM_005260380.3 |
XP_005260437.1
|
4, 5 |
520 |
|
XM_005260381.3 |
XP_005260438.1
|
4, 5 |
520 |
|
XM_005260382.4 |
XP_005260439.1
|
4, 5 |
520 |
|
XM_011528756.2 |
XP_011527058.1
|
3, 4 |
406 |
|
XM_011528757.2 |
XP_011527059.1
|
4, 5 |
354 |
|
GenBank mRNA |
AF149310.1 |
- |
4, 5 |
442 |
|
AF169385.1 |
- |
4, 5 |
517 |
|
BC033591.1 |
- |
3, 4 |
350 |
5167214 |
View Full Product Details