Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001126103.2 | NP_001119575.1 | 5 | 476 | |
NM_001126104.2 | NP_001119576.1 | 3 | 356 | ||
NM_001319999.1 | NP_001306928.1 | 3 | 286 | ||
NM_001320000.1 | NP_001306929.1 | 4 | 360 | ||
NM_001320001.1 | NP_001306930.1 | 3 | 352 | ||
NM_001320002.1 | NP_001306931.1 | 3 | 311 | ||
NM_001320003.1 | NP_001306932.1 | 3 | 282 | ||
NM_001320004.1 | NP_001306933.1 | 3 | 233 | ||
NM_001320005.1 | NP_001306934.1 | 2 | 197 | ||
NM_001320006.1 | NP_001306935.1 | 4 | 356 | ||
NM_013277.4 | NP_037409.2 | 5 | 465 | ||
XM_006719359.1 | XP_006719422.1 | 3 | 184 | ||
XM_011538238.1 | XP_011536540.1 | 3 | 141 | ||
XM_017019220.1 | XP_016874709.1 | 3 | 458 | ||
GenBank mRNA | AB030251.1 | - | 3 | 192 | |
AK000733.1 | - | 3 | 199 | ||
AK225901.1 | - | 2 | 127 | ||
AK315785.1 | - | 5 | 413 | ||
AL136794.1 | - | 5 | 346 | ||
BC024144.2 | - | 3 | 177 | ||
BC032754.1 | - | 3 | 183 | 5583315 | |
BU940828.1 | - | 4 | 250 | ||
CR533565.1 | - | 2 | 122 | ||
DA051287.1 | - | 3 | 190 | ||
DA441098.1 | - | 3 | 254 | ||
DA502516.1 | - | 3 | 168 | ||
DA812983.1 | - | 4 | 264 | ||
DB076571.1 | - | 3 | 253 | ||
DB084617.1 | - | 3 | 234 | ||
DB090068.1 | - | 2 | 134 | ||
DB164810.1 | - | 3 | 169 |