Gene Transcripts

Gene Symbol
ATG4B
Entrez Gene ID
23192
Gene Name
autophagy related 4B cysteine peptidase
Gene Aliases
APG4B, AUTL1
Chromosome Location
Chr. 2: 241637612 - 241673857 on Build GRCh38
UniGene ID
Hs.283610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013325.4 NP_037457.3 13 2802
NM_178326.2 NP_847896.1 13 2822
XM_005246992.3 XP_005247049.2 12 3573
XM_005246993.2 XP_005247050.2 13 2824
GenBank mRNA AB023160.2 - 13 2705
AK027332.1 - 11 1078
AK027763.1 - 13 2754
AK123425.1 - 4 2169
AK125277.1 - 13 3246
AL050288.1 - 1 1602
AL133638.1 - 1 903
BC000719.2 - 13 2708 3504267

View Full Product Details